Assay Details
Target Gene Details
Entrez Gene ID: | 13682 |
Gene Name: | eukaryotic translation initiation factor 4A2 |
Gene Aliases: |
4833432N07Rik, BM-010, Ddx2b, Eif4, eIF-4A-II, eIF4A-II |
Location: |
Chr.16:23107468-23114132 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eif4a2 | NM_001123037.2 | 1 | 176 | NP_001116509.1 |
NM_001123038.2 | 1 | 176 | NP_001116510.1 | |
NM_013506.3 | 1 | 176 | NP_038534.2 | |
NR_110335.1 | 1 | 176 | ||
BY286182.1 | 1 | 177 | ||
CF723811.1 |