Assay Details
Target Gene Details
Entrez Gene ID: | 385668 |
Gene Name: | Leber congenital amaurosis 5-like |
Gene Aliases: |
4921526F01Rik, Lcca5l |
Location: |
Chr.16:96158405-96193993 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lca5l | NM_001001492.2 | 5 | 732 | NP_001001492.2 |
AK029554.1 | 5 | 726 | BAC26513.1 | |
AK133316.1 | 5 | 732 | ||
BC141285.1 | 4 | 582 | ||
BC145611.1 | 4 | 582 |
Target Gene Details
Entrez Gene ID: | 71446 |
Gene Name: | tryptophan rich basic protein |
Gene Aliases: |
5530402J05Rik, C030018G21Rik, Chd5 |
Location: |
Chr.16:96145354-96189247 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Wrb |