Assay Details
Target Gene Details
Entrez Gene ID: | 83925 |
Gene Name: | trichorhinophalangeal syndrome I (human) |
Gene Aliases: |
AI115454, AI447310, D15Ertd586e |
Location: |
Chr.15:50654757-50890747 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Trps1 | NM_032000.2 | 3 | 2121 | NP_114389.2 |
AF346836.1 | 4 | 2046 | AAK39508.1 | |
AK142355.1 | 3 | 1791 | BAE25041.1 | |
BC037058.1 | AAH37058.1 | |||
BC049857.1 | AAH49857.1 | |||
BC083110.1 | 2 | 879 | AAH83110.1 | |
BC099962.1 | 3 | 2086 | AAH99962.1 |
More Information
Set Membership: |
Intragenic Exonic |