Assay Details
Target Gene Details
Entrez Gene ID: | 11911 |
Gene Name: | activating transcription factor 4 |
Gene Aliases: |
Atf-4, C/ATF, CREB2, TAXREB67 |
Location: |
Chr.15:80255184-80257545 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Atf4 | NM_001287180.1 | 4 | 833 | NP_001274109.1 |
NM_009716.3 | 3 | 1116 | NP_033846.2 | |
AB012277.1 | 1 | 264 | BAA25314.1 | |
AK003001.1 | 3 | 1120 | BAC25014.1 | |
AK138657.1 | 3 | 801 | BAE23736.1 | |
AK144777.1 | 3 | 802 | BAE26060.1 | |
AK146193.1 | 3 | 804 | BAE26967.1 | |
AK150231.1 | 3 | 803 | ||
AK155254.1 | 3 | 802 | BAE33149.1 | |
AK156298.1 | 3 | 796 | BAE33662.1 | |
AK177669.1 | 1 | 69 | ||
AK192031.1 | ||||
AK204555.1 | ||||
BC085169.1 | 3 | 1007 | AAH85169.1 | |
L13791.1 | 2 | 621 | AAA40476.1 | |
M94087.1 | 3 | 783 | AAA53043.1 | |
X61507.1 | 3 | 793 | CAA43723.1 |
More Information
Set Membership: |
Intragenic Exonic |