Assay Details
Target Gene Details
Entrez Gene ID: | 18163 |
Gene Name: | catenin (cadherin associated protein), delta 2 |
Gene Aliases: |
Catnd2, Nprap, neurojugin |
Location: |
Chr.15:30172593-31029344 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ctnnd2 | NM_008729.2 | 9 | 2021 | NP_032755.2 |
BC138260.1 | 9 | 1566 | ||
BC138261.1 | 9 | 1566 | ||
BC145217.1 | 9 | 1563 | ||
U90331.1 | 9 | 2024 | AAB82409.1 |
More Information
Set Membership: |
Intragenic Exonic |