Assay Details
Target Gene Details
Entrez Gene ID: | 16365 |
Gene Name: | immunoresponsive gene 1 |
Gene Aliases: |
AI323667, CAD |
Location: |
Chr.14:103047012-103056573 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Irg1 | NM_008392.1 | 5 | 2238 | NP_032418.1 |
AK152177.1 | 5 | 2237 | BAE31008.1 | |
AK152248.1 | 5 | 2237 | BAE31070.1 | |
AK152332.1 | 5 | 2237 | BAE31131.1 | |
AK152635.1 | 5 | 2237 | BAE31377.1 |
More Information
Set Membership: |
Intragenic Exonic |