Assay Details
Target Gene Details
Entrez Gene ID: | 110197 |
Gene Name: | diacylglycerol kinase, gamma |
Gene Aliases: |
2900055E17Rik, 90kDa, AI854428, Dagk3, E430001K23Rik, mKIAA4131 |
Location: |
Chr.16:22466568-22657299 on Build GRCm38 |
Assay Gene Location: | Within Exon 25 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dgkg | NM_138650.2 | 25 | 3194 | NP_619591.1 |
AK136842.1 | 1 | 147 | ||
AK220294.1 | 25 | 3136 | BAD90219.1 | |
BC015278.1 | 24 | 2767 | AAH15278.1 |
More Information
Set Membership: |
Intragenic Exonic |