Assay Details
Target Gene Details
Entrez Gene ID: | 239554 |
Gene Name: | FAD-dependent oxidoreductase domain containing 2 |
Gene Aliases: |
A430097D04Rik, D15Bwg0759e, Gm304 |
Location: |
Chr.15:77940522-77956771 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Foxred2 | NM_001017983.2 | 4 | 1094 | NP_001017983.2 |
NM_001168260.1 | 4 | 1192 | NP_001161732.1 | |
AK140027.1 | 4 | 1054 | BAE24215.1 | |
AK170795.1 | 4 | 1147 | BAE42033.1 | |
BC086662.1 | 4 | 1024 | AAH86662.1 |
More Information
Set Membership: |
Intragenic Exonic |