Assay Details
Target Gene Details
Entrez Gene ID: | 224273 |
Gene Name: | beta-gamma crystallin domain containing 3 |
Gene Aliases: |
5031404N19, AU022790 |
Location: |
Chr.16:59490775-59601399 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Crybg3 | NM_174848.3 | 19 | 4955 | NP_777273.2 |
AK030282.1 | 10 | 2399 | BAC26878.1 | |
AK162506.1 | 7 | 2790 | BAE36951.1 | |
BC043118.1 | 19 | 4955 | AAH43118.1 |
Target Gene Details
Entrez Gene ID: | 67014 |
Gene Name: | myc induced nuclear antigen |
Gene Aliases: |
1810047J07Rik, 2410057H13Rik, 3830408E23Rik, AI449204 |
Location: |
Chr.16:59471769-59492461 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mina | NM_025910.3 | 10 | 2062 | NP_080186.3 |
AK031671.1 | 10 | 1996 | BAC27503.1 | |
BC025109.1 | 10 | 1984 | AAH25109.1 |