Assay Details
Target Gene Details
Entrez Gene ID: | 387609 |
Gene Name: | zinc fingers and homeoboxes 2 |
Gene Aliases: |
Afr-1, Afr1, Raf, mKIAA0854 |
Location: |
Chr.15:57694667-57839832 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zhx2 | NM_199449.2 | 1 | 275 | NP_955520.1 |
AK031782.1 | 1 | 172 | BAC27546.1 | |
AK157631.1 | 1 | 262 | BAE34139.1 | |
BC059178.1 | AAH59178.1 |
More Information
Set Membership: |
Intragenic Exonic |