Assay Details
Target Gene Details
Entrez Gene ID: | 19270 |
Gene Name: | protein tyrosine phosphatase, receptor type, G |
Gene Aliases: |
5430405N12Rik, AW046354, AW549872, RPTPgamma |
Location: |
Chr.14:11553553-12242039 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptprg | NM_008981.3 | 1 | 522 | NP_033007.2 |
AK053271.1 | 1 | 494 | BAC35328.1 | |
L09562.1 | 1 | 63 | AAA40022.1 |
More Information
Set Membership: |
Intragenic Exonic |