Assay Details
Target Gene Details
Entrez Gene ID: | 12846 |
Gene Name: | catechol-O-methyltransferase |
Gene Aliases: |
Comt1, D16Wsu103e, D330014B15Rik |
Location: |
Chr.16:18406882-18426716 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Comt | NM_001111062.1 | 6 | 1213 | NP_001104532.1 |
NM_001111063.1 | 5 | 1024 | NP_001104533.1 | |
NM_007744.3 | 5 | 1156 | NP_031770.2 | |
AF076156.1 | 5 | 958 | AAC33334.1 | |
AK148311.1 | 6 | 880 | BAE28473.1 | |
AK188838.1 | 1 | 108 | ||
AK205701.1 | 1 | 146 | ||
AK206856.1 | 1 | 80 | ||
AK216466.1 | 1 | 85 | ||
BC010402.1 | 5 | 950 | AAH10402.1 |
More Information
Set Membership: |
Intragenic Exonic |