Assay Details
Target Gene Details
Entrez Gene ID: | 18155 |
Gene Name: | prepronociceptin |
Gene Aliases: |
N/OFQ, N23K, Npnc1, OFQ/N |
Location: |
Chr.14:65400673-65425581 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pnoc | NM_001205075.1 | 4 | 1119 | NP_001192004.1 |
NM_010932.2 | 4 | 1175 | NP_035062.1 | |
AK045893.1 | 4 | 912 | BAC32523.1 | |
BC119372.1 | 4 | 863 | ||
BC144865.1 | 4 | 807 | ||
D50055.1 | 4 | 802 | BAA08773.1 | |
D50056.1 | 4 | 872 | BAA08774.1 | |
D82866.1 | 4 | 1175 | BAA11614.1 |
More Information
Set Membership: |
Intragenic Exonic |