Assay Details
Target Gene Details
Entrez Gene ID: | 66609 |
Gene Name: | crystallin, zeta (quinone reductase)-like 1 |
Gene Aliases: |
2210407J23Rik, 2410006O11Rik, QOH-1 |
Location: |
Chr.16:91689322-91728802 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cryzl1 | NM_026994.1 | NP_081270.1 | ||
NM_133679.2 | NP_598440.1 | |||
AK008846.1 | ||||
AK010433.1 | BAB26935.1 | |||
AK017995.1 | ||||
AK040530.1 | 1 | 314 | ||
AK145827.1 | BAE26679.1 | |||
BC010479.1 | AAH10479.1 | |||
BY717916.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |