Assay Details
Target Gene Details
Entrez Gene ID: | 73225 |
Gene Name: | family with sequence similarity 118, member A |
Gene Aliases: |
- |
Location: |
Chr.15:85037015-85116933 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fam118a | NM_133750.4 | 9 | 2101 | NP_598511.1 |
AK014192.1 | 1 | 482 | ||
AK143690.1 | 9 | 2074 | BAE25500.1 | |
BC016425.1 | 9 | 1959 | AAH16425.1 | |
BC058794.1 | 9 | 2162 | AAH58794.1 | |
BC066174.1 | 9 | 2017 | AAH66174.1 |