Assay Details
Target Gene Details
Entrez Gene ID: | 73826 |
Gene Name: | polymerase (DNA-directed), delta interacting protein 3 |
Gene Aliases: |
1110008P04Rik, AA408269, AL022852, C77954, PDIP46, SKAR, mKIAA1649 |
Location: |
Chr.15:83125976-83149373 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Poldip3 | NM_178627.3 | 9 | 1525 | NP_848742.1 |
AK081139.1 | 9 | 1485 | BAC38144.1 | |
AK083924.1 | 9 | 1526 | BAC39068.1 | |
AK087446.1 | 9 | 1534 | BAC39877.1 | |
AK088088.1 | 5 | 747 | ||
AK150130.1 | 8 | 1416 | BAE29328.1 | |
AK152060.1 | 9 | 1566 | BAE30916.1 | |
AK167552.1 | 9 | 1469 | BAE39617.1 | |
AK184858.1 | 1 | 57 | ||
AK220221.1 | 9 | 1541 | BAD90146.1 | |
BC037652.1 | 9 | 1510 | AAH37652.2 | |
BC046505.1 | 9 | 1454 | AAH46505.2 | |
BC058225.1 | 9 | 1521 | AAH58225.2 |
More Information
Set Membership: |
Intragenic Exonic |