Assay Details
Target Gene Details
Entrez Gene ID: | 69089 |
Gene Name: | oxidase assembly 1-like |
Gene Aliases: |
1810020M02Rik, AI894287 |
Location: |
Chr.14:54360841-54369669 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Oxa1l | NM_026936.3 | 10 | 2385 | NP_081212.1 |
AK052432.1 | 10 | 2386 | BAC34985.1 | |
AK154532.1 | 10 | 2386 | BAE32657.1 |
Target Gene Details
Entrez Gene ID: | 20540 |
Gene Name: | solute carrier family 7 (cationic amino acid transporter, y+ system), member 7 |
Gene Aliases: |
AI790233, my+lat1 |
Location: |
Chr.14:54369442-54417702 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc7a7 | NM_001253679.1 | 11 | 2048 | NP_001240608.1 |
NM_001253680.1 | 10 | 2066 | NP_001240609.1 | |
NM_011405.4 | 12 | 2205 | NP_035535.2 | |
AJ012754.1 | 10 | 2056 | CAA10170.1 | |
AJ130943.1 | 11 | 2048 | CAA10255.1 | |
AK150950.1 | 12 | 2205 | BAE29983.1 | |
AK151718.1 | 12 | 2205 | BAE30637.1 | |
AK152637.1 | 10 | 1971 | BAE31379.1 | |
AK170851.1 | 10 | 1977 | BAE42073.1 | |
AV487330.1 | ||||
BC014709.1 | 10 | 2054 | AAH14709.1 |
More Information
Set Membership: |
Intragenic Exonic |