Assay Details
Target Gene Details
Entrez Gene ID: | 106347 |
Gene Name: | immunoglobulin-like domain containing receptor 1 |
Gene Aliases: |
AU041483, AU044638 |
Location: |
Chr.16:36693978-36726804 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ildr1 | NM_001285788.1 | 7 | 1336 | NP_001272717.1 |
NM_001285791.1 | 6 | 1204 | NP_001272720.1 | |
NM_134109.2 | 7 | 1336 | NP_598870.1 | |
AK035504.1 | 7 | 1265 | BAC29081.1 | |
AK036844.1 | 7 | 1262 | BAC29604.1 | |
AK136284.1 | 7 | 1336 | BAE22913.1 | |
BC010485.1 | 7 | 1276 | AAH10485.1 | |
BC057644.1 | 6 | 1201 | AAH57644.1 |
More Information
Set Membership: |
Intragenic Exonic |