Assay Details
Target Gene Details
Entrez Gene ID: | 13837 |
Gene Name: | Eph receptor A3 |
Gene Aliases: |
AW492086, Cek4, ETK1, End3, Hek, Hek4, Mek4, Tyro4 |
Location: |
Chr.16:63545218-63864157 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Epha3 | NM_010140.3 | 3 | 814 | NP_034270.1 |
AK041935.1 | 3 | 815 | BAC31104.1 | |
AK045219.1 | 3 | 815 | BAC32266.1 | |
AK084921.1 | 3 | 794 | BAC39310.1 | |
AK132335.1 | 3 | 763 | BAE21112.1 | |
BC138641.1 | 3 | 737 | ||
BC138643.1 | 3 | 737 | ||
BC145349.1 | 3 | 737 | ||
M68513.1 | 3 | 792 | AAA39521.1 | |
M68515.1 | 3 | 792 | AAA39522.1 |
More Information
Set Membership: |
Intragenic Exonic |