Assay Details
Target Gene Details
Entrez Gene ID: | 71704 |
Gene Name: | Rho guanine nucleotide exchange factor (GEF) 3 |
Gene Aliases: |
1200004I24Rik, 9830169H03Rik, C76747, Xpln |
Location: |
Chr.14:27114823-27403916 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Arhgef3 | NM_001289686.1 | 10 | 921 | NP_001276615.1 |
NM_001289687.1 | 7 | 792 | NP_001276616.1 | |
NM_001289688.1 | 7 | 966 | NP_001276617.1 | |
NM_027871.2 | 8 | 766 | NP_082147.1 | |
AK004600.2 | 8 | 737 | BAB23401.1 | |
AK029872.1 | 11 | 951 | BAC26652.1 | |
BC005517.1 | 7 | 763 | AAH05517.1 | |
BC007153.1 | AAH07153.1 | |||
BC012262.1 | 7 | 744 | AAH12262.1 |
More Information
Set Membership: |
Intragenic Exonic |