Assay Details
Target Gene Details
Entrez Gene ID: | 68750 |
Gene Name: | ras responsive element binding protein 1 |
Gene Aliases: |
1110037N09Rik, AA414966, B930013M22Rik |
Location: |
Chr.13:37778382-37952005 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rreb1 | NM_001177869.1 | 11 | 4770 | NP_001171340.1 |
NR_033218.1 | 11 | 6069 | ||
NR_033615.1 | 9 | 2764 | ||
AK147260.1 | 11 | 6070 | BAE27803.1 | |
AK147653.1 | 11 | 6070 | BAE28051.1 | |
AY946044.1 | 9 | 4461 | AAX83010.1 |