Assay Details
Target Gene Details
Entrez Gene ID: | 11843 |
Gene Name: | ADP-ribosylation factor 4 |
Gene Aliases: |
AA407803 |
Location: |
Chr.14:26638197-26657258 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Arf4 | NM_007479.3 | 6 | 906 | NP_031505.1 |
AK002302.1 | 6 | 929 | BAB21999.1 | |
AK013892.1 | 6 | 961 | BAB29041.1 | |
AK081686.1 | 6 | 906 | BAC38292.1 | |
AK153011.1 | 6 | 890 | BAE31650.1 | |
AK168793.1 | 6 | 927 | BAE40626.1 | |
AK206349.1 | ||||
AK211971.1 | 1 | 53 | ||
BC115650.1 | 6 | 785 | ||
BC115651.1 | 6 | 785 | ||
D87901.1 | 6 | 723 | BAA13493.1 |
More Information
Set Membership: |
Intragenic Exonic |