Assay Details
Target Gene Details
Entrez Gene ID: | 627191 |
Gene Name: | synapse differentiation inducing 1 like |
Gene Aliases: |
DSPC1, Gm261, Tmem90a |
Location: |
Chr.12:84677277-84698907 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Syndig1l | NM_001033334.2 | 1 | 141 | NP_001028506.1 |
AK140195.1 | 1 | 141 | BAE24274.1 | |
AY993933.1 | 1 | 88 | AAY42815.1 | |
AY993934.1 | 1 | 165 | AAY42816.1 |