Assay Details
Target Gene Details
Entrez Gene ID: | 56784 |
Gene Name: | Ral GTPase activating protein, alpha subunit 1 |
Gene Aliases: |
2310003F20Rik, 4930400K19Rik, AI563624, GRIPE, Garnl1, Tulip1, mKIAA0884 |
Location: |
Chr.12:55602890-55821575 on Build GRCm38 |
Assay Gene Location: | Within Exon 43 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ralgapa1 | NM_001003719.2 | 41 | 7533 | NP_001003719.1 |
NM_001112714.2 | 41 | 7561 | NP_001106185.1 | |
NM_001286263.1 | 43 | 7705 | NP_001273192.1 | |
NM_019994.5 | 42 | 7564 | NP_064378.4 | |
AK009111.1 | 3 | 792 | ||
AY066011.1 | 29 | 5162 | AAL47577.1 | |
AY596972.1 | 42 | 7215 | AAT49273.1 | |
AY596973.1 | 41 | 7212 | AAT49274.1 | |
AY596974.1 | 41 | 7184 | AAT49275.1 | |
CD776675.1 | 1 | 717 |
More Information
Set Membership: |
Intragenic Exonic |