Assay Details
Target Gene Details
Entrez Gene ID: | 11774 |
Gene Name: | adaptor-related protein complex 3, beta 1 subunit |
Gene Aliases: |
AP-3, AU015684, C78395, Hps2, beta3A, pe, pearl, rim2 |
Location: |
Chr.13:94358952-94566316 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ap3b1 | NM_009680.3 | 1 | 138 | NP_033810.2 |
AF103809.2 | 1 | 75 | AAC78338.1 | |
AK143559.1 | 1 | 134 | BAE25435.1 | |
AK163957.1 | 1 | 76 | BAE37552.1 | |
AK171595.1 | 1 | 139 | ||
BC015068.1 | 1 | 136 | AAH15068.1 |
More Information
Set Membership: |
Intragenic Exonic |