Assay Details
Target Gene Details
Entrez Gene ID: | 328232 |
Gene Name: | glucose-fructose oxidoreductase domain containing 1 |
Gene Aliases: |
9630032O13, AI850995 |
Location: |
Chr.13:43195240-43304172 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gfod1 | NM_001033399.4 | 1 | 624 | NP_001028571.1 |
AK147458.1 | 1 | 625 | BAE27925.1 | |
BC139026.1 | 1 | 444 | ||
BC139027.1 | 1 | 444 |
More Information
Set Membership: |
Intragenic Exonic |