Assay Details
Target Gene Details
Entrez Gene ID: | 94253 |
Gene Name: | HECT, C2 and WW domain containing E3 ubiquitin protein ligase 1 |
Gene Aliases: |
9330116H24Rik, AV273951, E130207I19Rik, Nedl1 |
Location: |
Chr.13:14226438-14523280 on Build GRCm38 |
Assay Gene Location: | Within Exon 29 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hecw1 | NM_001081348.3 | 29 | 5857 | NP_001074817.3 |
AK031094.1 | 1 | 492 | ||
AK033922.1 | 9 | 1792 | BAC28516.1 | |
AK164878.1 | 1 | 497 | ||
AK172932.1 | 20 | 4142 | BAD32210.1 |
More Information
Set Membership: |
Intragenic Exonic |