Assay Details
Target Gene Details
Entrez Gene ID: | 69635 |
Gene Name: | death associated protein kinase 1 |
Gene Aliases: |
D13Ucla1, DAP-Kinase |
Location: |
Chr.13:60601947-60763191 on Build GRCm38 |
Assay Gene Location: | Within Exon 27 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dapk1 | NM_001285917.1 | NP_001272846.1 | ||
NM_029653.3 | 26 | 4611 | NP_083929.2 | |
NM_134062.2 | NP_598823.1 | |||
AK013153.1 | BAB28681.1 | |||
AK145922.1 | BAE26753.1 | |||
AY245540.2 | AAO91934.2 | |||
AY245541.1 | 26 | 4597 | AAO91935.1 | |
BC021490.1 | AAH21490.1 | |||
BC026671.1 | 1 | 1251 | AAH26671.1 | |
X97048.1 | 25 | 4446 | CAA65762.1 |
More Information
Set Membership: |
Intragenic Exonic |