Assay Details
Target Gene Details
Entrez Gene ID: | 26914 |
Gene Name: | H2A histone family, member Y |
Gene Aliases: |
H2AF12M, mH2a1, macroH2A1 |
Location: |
Chr.13:56073622-56135626 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
H2afy | NM_001159513.1 | 9 | 1710 | NP_001152985.1 |
NM_001159514.1 | 9 | 1704 | NP_001152986.1 | |
NM_001159515.1 | 9 | 1701 | NP_001152987.1 | |
NM_012015.2 | 9 | 1713 | NP_036145.1 | |
AK003531.1 | 1 | 481 | BAB22841.1 | |
AK212046.1 | 1 | 35 | ||
BE988321.1 | 1 | 266 |
More Information
Set Membership: |
Intragenic Exonic |