Assay Details
Target Gene Details
Entrez Gene ID: | 17300 |
Gene Name: | forkhead box C1 |
Gene Aliases: |
FREAC3, Fkh1, Mf1, Mf4, ch, fkh-1, frkhda |
Location: |
Chr.13:31806646-31810635 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Foxc1 | NM_008592.2 | 1 | 3328 | NP_032618.2 |
AF045017.1 | 1 | 3314 | AAC24209.1 |
More Information
Set Membership: |
Intragenic Exonic |