Assay Details
Target Gene Details
Entrez Gene ID: | 17933 |
Gene Name: | myelin transcription factor 1-like |
Gene Aliases: |
2900046C06Rik, 2900093J19Rik, C630034G21Rik, Nztf1, Pmng1, Png-1, mKIAA1106 |
Location: |
Chr.12:29528311-29923216 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Myt1l | NM_001093775.1 | 10 | 1908 | NP_001087244.1 |
NM_001093776.1 | 10 | 1820 | NP_001087245.1 | |
NM_001093778.1 | 10 | 1905 | NP_001087247.1 | |
NM_008666.3 | 10 | 1908 | NP_032692.2 | |
AB093283.1 | 7 | 1267 | BAC41467.1 | |
AB212898.1 | 9 | 1506 | BAE48255.1 | |
AF004295.1 | 10 | 1727 | AAC53457.1 | |
BC094438.1 | 6 | 1145 | ||
BC131677.1 | 9 | 1509 | ||
CD802997.1 | ||||
CF536651.1 | ||||
U86338.1 | 10 | 1630 | AAC53157.1 |
More Information
Set Membership: |
Intragenic Exonic |