Assay Details
Target Gene Details
Entrez Gene ID: | 68018 |
Gene Name: | collagen, type IV, alpha 3 (Goodpasture antigen) binding protein |
Gene Aliases: |
2810404O15Rik, 9230101K08Rik, AU016711, CERT, GPBP |
Location: |
Chr.13:96542735-96640167 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Col4a3bp | NM_001164222.1 | 1 | 352 | NP_001157694.1 |
NM_023420.2 | 1 | 352 | NP_075909.1 | |
AF232932.1 | 1 | 366 | AAG42048.1 | |
AF232934.1 | 1 | 366 | AAG42050.1 | |
AK012989.1 | 1 | 333 | BAB28581.1 | |
AK030305.1 | 2 | 260 | BAC26888.1 | |
AK038061.1 | 1 | 342 | BAC29927.1 | |
AK038803.1 | 1 | 333 | ||
AK047100.1 | 1 | 349 | BAC32962.1 | |
AK080958.1 | 1 | 350 | BAC38095.2 | |
AK163624.1 | 1 | 342 | BAE37427.1 | |
BB654693.1 | 1 | 353 | ||
BC016197.1 | 1 | 315 | AAH16197.1 |
More Information
Set Membership: |
Intragenic Exonic |