Assay Details
Target Gene Details
Entrez Gene ID: | 380840 |
Gene Name: | LYR motif containing 4 |
Gene Aliases: |
BC034664, Gm903 |
Location: |
Chr.13:35978797-36117357 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lyrm4 | NM_201358.2 | 3 | 961 | NP_958746.1 |
AK050553.1 | 3 | 936 | BAC34321.1 |