Assay Details
Target Gene Details
Entrez Gene ID: | 373852 |
Gene Name: | RIKEN cDNA 4833422C13 gene |
Gene Aliases: |
- |
Location: |
Chr.13:91701665-91741872 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
4833422C13Rik | NR_015501.1 | 2 | 2449 | |
AK048302.1 | 2 | 2450 |
Target Gene Details
Entrez Gene ID: | 66970 |
Gene Name: | single-stranded DNA binding protein 2 |
Gene Aliases: |
1500004K09Rik, 2310079I02Rik, 9330163K02Rik, A830008M03Rik, AU067692, AW558684, Hspc116, SSDP2 |
Location: |
Chr.13:91460979-91706175 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ssbp2 | NM_024186.5 | 16 | 2435 | NP_077148.1 |
NM_024272.4 | 17 | 2525 | NP_077234.2 | |
AK133699.1 | 16 | 2380 | ||
AY037837.1 | 17 | 2481 | AAK71881.1 |
More Information
Set Membership: |
Intragenic Exonic |