Assay Details
Target Gene Details
Entrez Gene ID: | 23849 |
Gene Name: | Kruppel-like factor 6 |
Gene Aliases: |
AI448727, BCD1, C86813, CPBP, Copeb, FM2, FM6, Ierepo1, Ierepo3, R75280, Zf9 |
Location: |
Chr.13:5861489-5870393 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Klf6 | NM_011803.2 | 4 | 3682 | NP_035933.2 |
AK144137.1 | BAE25722.1 | |||
AK146615.1 | 4 | 3633 | BAE27304.1 | |
AK147904.1 | 4 | 3659 | BAE28217.1 | |
AK151769.1 | 4 | 3504 | BAE30675.1 | |
AK159420.1 | 4 | 3540 | BAE35068.1 |
More Information
Set Membership: |
Intragenic Exonic |