Assay Details
Target Gene Details
Entrez Gene ID: | 74338 |
Gene Name: | solute carrier family 6 (neurotransmitter transporter), member 19 |
Gene Aliases: |
4632401C08Rik, B<0>AT1 |
Location: |
Chr.13:73679745-73700695 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc6a19 | NM_028878.3 | 12 | 1879 | NP_083154.1 |
AB096860.1 | 10 | 1562 | BAC66209.1 | |
AJ633679.1 | 12 | 1888 | CAG17898.1 | |
AK014544.1 | 12 | 1880 | BAB29422.1 | |
AK028784.1 | 12 | 1880 | BAC26119.1 | |
AK128923.1 | 11 | 2222 | ||
AK162399.1 | 11 | 1624 | BAE36894.1 | |
BC107411.1 | 12 | 1880 | AAI07412.1 | |
BC107412.2 | 12 | 1884 | AAI07413.1 |
More Information
Set Membership: |
Intragenic Exonic |