Assay Details
Target Gene Details
Entrez Gene ID: | 20354 |
Gene Name: | sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4D |
Gene Aliases: |
CD100, Semacl2, Semaj, Semcl2, coll-4 |
Location: |
Chr.13:51685522-51793747 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sema4d | NM_001281880.1 | 17 | 4157 | NP_001268809.1 |
NM_013660.4 | 16 | 4093 | NP_038688.2 | |
AK088653.1 | 13 | 3375 | BAC40480.1 | |
AK132688.1 | 17 | 4083 | BAE21302.1 | |
AK150911.1 | 15 | 3985 | ||
BC049780.1 | 16 | 3990 | AAH49780.2 | |
BE692125.1 |
More Information
Set Membership: |
Intragenic Exonic |