Assay Details
Target Gene Details
Entrez Gene ID: | 18976 |
Gene Name: | pro-opiomelanocortin-alpha |
Gene Aliases: |
ACTH, BE, Beta-LPH, Clip, Gamma-LPH, Npp, Pomc-1, Pomc1, alpha-MSH, alphaMSH, beta-MSH, gamma-MSH |
Location: |
Chr.12:3954945-3960643 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pomc | NM_001278581.1 | 4 | 582 | NP_001265510.1 |
NM_001278582.1 | 4 | 517 | NP_001265511.1 | |
NM_001278583.1 | 4 | 487 | NP_001265512.1 | |
NM_001278584.1 | 3 | 376 | NP_001265513.1 | |
NM_008895.4 | 3 | 406 | NP_032921.1 | |
AK017492.1 | 3 | 402 | BAB30771.1 | |
AK017581.1 | 3 | 371 | BAB30818.1 | |
AK030714.1 | 4 | 481 | BAC27095.1 | |
AK077426.1 | 3 | 370 | BAC36795.1 | |
AK133592.1 | 3 | 401 | BAE21739.1 | |
AK133646.1 | 3 | 401 | BAE21764.1 | |
AK133740.1 | 3 | 400 | BAE21815.1 | |
AK133776.1 | 3 | 402 | BAE21833.1 | |
AK133800.1 | 3 | 402 | BAE21850.1 | |
AW120866.1 | ||||
BB022229.2 | 1 | 62 | ||
BC061215.1 | 3 | 400 | AAH61215.1 |