Assay Details
Target Gene Details
Entrez Gene ID: | 76357 |
Gene Name: | TRM5 tRNA methyltransferase 5 |
Gene Aliases: |
2610027O18Rik, mKIAA1393 |
Location: |
Chr.12:73268515-73286787 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Trmt5 | NM_029580.3 | 4 | 902 | NP_083856.1 |
AK011576.1 | 4 | 903 | BAB27710.1 | |
AK049127.1 | 4 | 901 | ||
AK220389.1 | 3 | 649 | BAD90443.1 | |
BC012521.1 | 4 | 903 | AAH12521.1 |
More Information
Set Membership: |
Intragenic Exonic |