Assay Details
Target Gene Details
Entrez Gene ID: | 70059 |
Gene Name: | degenerative spermatocyte homolog 2 (Drosophila), lipid desaturase |
Gene Aliases: |
2210008A03Rik, AI852933, DES2 |
Location: |
Chr.12:108686792-108702306 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Degs2 | NM_001171002.1 | 2 | 398 | NP_001164473.1 |
NM_027299.5 | 2 | 398 | NP_081575.2 | |
AF466377.1 | 2 | 386 | AAM12533.1 | |
AK008666.1 | 2 | 399 | ||
AK162979.1 | 2 | 391 | BAE37145.1 | |
BC016427.1 | 2 | 380 |
More Information
Set Membership: |
Intragenic Exonic |