Assay Details
Target Gene Details
Entrez Gene ID: | 70357 |
Gene Name: | Kv channel-interacting protein 1 |
Gene Aliases: |
KCHIP1, Kchip1.2 |
Location: |
Chr.11:33629339-33993193 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kcnip1 | NM_001190885.1 | 9 | 1716 | NP_001177814.1 |
NM_001190886.1 | 8 | 1672 | NP_001177815.1 | |
NM_001290690.1 | 8 | 1871 | NP_001277619.1 | |
NM_027398.3 | 8 | 1683 | NP_081674.2 | |
AK014313.1 | 8 | 1671 | ||
AK045948.1 | 9 | 1717 | BAC32543.1 | |
AK046398.1 | 9 | 1705 | BAC32705.1 | |
AK081845.1 | 8 | 1673 | BAC38347.1 | |
AY171234.1 | 8 | 1747 | AAN77492.1 | |
BC034241.1 | 8 | 1625 | AAH34241.1 | |
DQ148493.1 | 8 | 1625 | AAZ77810.1 | |
DQ148494.1 | 8 | 1673 | AAZ77811.1 |