Assay Details
Target Gene Details
Entrez Gene ID: | 14784 |
Gene Name: | growth factor receptor bound protein 2 |
Gene Aliases: |
AA408164, Ash |
Location: |
Chr.11:115644130-115708597 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Grb2 | NM_008163.3 | 6 | 1406 | NP_032189.1 |
AK077750.1 | 5 | 1226 | ||
AK079087.1 | 5 | 1234 | ||
AK148148.1 | 6 | 1371 | BAE28376.1 | |
AK155800.1 | 6 | 1361 | BAE33439.1 | |
AK166142.1 | 6 | 1360 | BAE38595.1 | |
AK170845.1 | 5 | 1197 | BAE42068.1 | |
AK172002.1 | 6 | 1382 | ||
AK184348.1 | ||||
AK192421.1 | ||||
AK194267.1 | ||||
AK195957.1 | ||||
BC052377.1 | 6 | 1406 | AAH52377.1 | |
BC085254.1 | 6 | 1242 | AAH85254.1 | |
D85748.1 | 4 | 1030 | BAA12862.1 | |
U07617.1 | 5 | 1155 | AAB40022.1 |