Assay Details
Target Gene Details
Entrez Gene ID: | 53860 |
Gene Name: | septin 9 |
Gene Aliases: |
MSF1, Msf, PNUTL4, Sint1 |
Location: |
Chr.11:117199661-117362325 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sept9 | NM_001113486.1 | 12 | 2664 | NP_001106958.1 |
NM_001113487.1 | 11 | 2844 | NP_001106959.1 | |
NM_001113488.1 | 10 | 2013 | NP_001106960.1 | |
NM_017380.2 | 10 | 1994 | NP_059076.1 | |
AJ250723.1 | 10 | 2000 | CAB59833.1 | |
AK031757.1 | 10 | 1889 | BAC27538.1 | |
AK122415.2 | 11 | 2638 | BAC65697.2 | |
AK141312.1 | 12 | 2605 | BAE24646.1 | |
AK164581.1 | 9 | 4608 | ||
BC046524.1 | 12 | 2664 | AAH46524.1 |
More Information
Set Membership: |
Intragenic Exonic |