Assay Details
Target Gene Details
Entrez Gene ID: | 83813 |
Gene Name: | tyrosine kinase, non-receptor, 1 |
Gene Aliases: |
Kos1, Tnk1a, Tnk1b |
Location: |
Chr.11:69850563-69858730 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tnk1 | NM_031880.3 | 12 | 1769 | NP_114086.3 |
AF332512.1 | 10 | 1589 | AAK35164.1 | |
AK033440.1 | 7 | 1140 | BAC28288.1 | |
AK076220.1 | 12 | 1776 | ||
BC029623.1 | 11 | 1756 | AAH29623.1 | |
BC055303.1 | 11 | 1673 | AAH55303.1 |