Assay Details
Target Gene Details
Entrez Gene ID: | 53860 |
Gene Name: | septin 9 |
Gene Aliases: |
MSF1, Msf, PNUTL4, Sint1 |
Location: |
Chr.11:117199661-117362325 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sept9 | NM_001113486.1 | 3 | 597 | NP_001106958.1 |
NM_001113487.1 | 2 | 777 | NP_001106959.1 | |
AK122415.2 | 2 | 571 | BAC65697.2 | |
AK141312.1 | 3 | 538 | BAE24646.1 | |
BC046524.1 | 3 | 597 | AAH46524.1 | |
BU148654.1 |
More Information
Set Membership: |
Intragenic Exonic |