Assay Details
Target Gene Details
Entrez Gene ID: | 53860 |
Gene Name: | septin 9 |
Gene Aliases: |
MSF1, Msf, PNUTL4, Sint1 |
Location: |
Chr.11:117199661-117362325 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sept9 | NM_001113486.1 | NP_001106958.1 | ||
NM_001113487.1 | 1 | 108 | NP_001106959.1 | |
AK141312.1 | BAE24646.1 | |||
BC046524.1 | AAH46524.1 | |||
BU148654.1 | 1 | 108 |
More Information
Set Membership: |
Intragenic Exonic |