Assay Details
Target Gene Details
Entrez Gene ID: | 632687 |
Gene Name: | membrane-associated ring finger (C3HC4) 10 |
Gene Aliases: |
4933417C16Rik, AV039223, OTTMUSG00000002847, Rnf190 |
Location: |
Chr.11:105360798-105456863 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
March10 | NM_001039242.2 | 1 | 173 | NP_001034331.1 |
NM_172568.2 | 1 | 173 | NP_766156.2 | |
AK132734.1 | 1 | 162 | BAE21325.1 | |
BC100474.1 | 1 | 166 | AAI00475.1 | |
BY099161.1 | 1 | 174 |
More Information
Set Membership: |
Intragenic Exonic |