Assay Details
Target Gene Details
Entrez Gene ID: | 21428 |
Gene Name: | MAX-like protein X |
Gene Aliases: |
Tcfl4, bHLHd13 |
Location: |
Chr.11:101086709-101092456 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mlx |
Target Gene Details
Entrez Gene ID: | 19183 |
Gene Name: | proteasome (prosome, macropain) 26S subunit, ATPase 3, interacting protein |
Gene Aliases: |
C79099, GT198, HOP2, Tbpip |
Location: |
Chr.11:101092141-101095435 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Psmc3ip | NM_008949.3 | 8 | 701 | NP_032975.1 |
AB000121.1 | 8 | 666 | BAA23155.1 | |
AK133462.1 | 8 | 669 | BAE21670.1 | |
BC030169.1 | 8 | 678 | AAH30169.1 | |
BF658859.1 | 1 | 284 | ||
FJ937966.1 | ||||
FJ937967.1 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic |