Assay Details
Target Gene Details
Entrez Gene ID: | 57230 |
Gene Name: | SAP30 binding protein |
Gene Aliases: |
2700016D05Rik, Hcngp |
Location: |
Chr.11:115933578-115965534 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sap30bp | NM_020483.3 | 11 | 1425 | NP_065229.2 |
AK147869.1 | BAE28192.1 | |||
BC021757.1 | 11 | 1425 | AAH21757.1 |
More Information
Set Membership: |
Intragenic Exonic |