Assay Details
Target Gene Details
Entrez Gene ID: | 380732 |
Gene Name: | mast cell immunoglobulin like receptor 1 |
Gene Aliases: |
Allergin-1, Gm885, Mca32 |
Location: |
Chr.11:106750652-106768794 on Build GRCm38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Milr1 | NM_001271375.1 | NP_001258304.1 | ||
NR_073165.1 | ||||
AK137500.1 | BAE23382.1 | |||
AK169925.1 | BAE41463.1 |
Target Gene Details
Entrez Gene ID: | 50776 |
Gene Name: | polymerase (DNA directed), gamma 2, accessory subunit |
Gene Aliases: |
- |
Location: |
Chr.11:106754821-106780010 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Polg2 | NM_015810.2 | 8 | 1452 | NP_056625.2 |
NR_027785.1 | 8 | 1305 | ||
AF177202.1 | AAD56641.1 | |||
AK041591.1 | 8 | 1424 | BAE20595.1 | |
AK169977.1 | 8 | 1694 | ||
BC119131.1 | 8 | 1381 | ||
BE853186.1 | 1 | 134 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic |